Please login to your online account to display your discounted pricing

Human IGF-I Quantikine ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSSG100

 View more versions of this product

Catalog No. SG100

  • / Each
 

Description & Specifications

Specifications

Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma
Packaging Quantity 6 Plates
Quantity 1 Pk.
Sample Volume Cell Culture Supernates (50μL), Serum (20μL), EDTA Plasma (20μL), Heparin Plasma (20μL)
Sufficient For Sufficient materials to run ELISAs on at least six 96-well plates
Gene ID (Entrez) 3479
Sensitivity 0.056ng/mL
Species Human
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules