Please login to your online account to display your discounted pricing

eBioscience Human IgE Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human IgE.

$665.55 - $4,197.75

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym Immunoglobulin E
Target IgE
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IgE

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125321

 
ebioscience
LSBMS2097CE
96 tests Each for $665.55

501125323

 
ebioscience
LSBMS2097TENCE
10 x 96 tests Each for $4,197.75
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym Immunoglobulin E
Target IgE
Conjugate HRP
Sample Type Serum, plasma
Sensitivity 0.5ng/mL
Storage Requirements 2° to 8°C
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.