missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Human IgE ELISA Ready-SET-Go!™ Kit

For quantitative detection of human IgE.

Manufacturer:  Invitrogen™ LS885061076

 View more versions of this product

Catalog No. 501125152



Includes: Capture Antibody Concentrate
Detection Antibody Concentrate
human IgE isotype control (standard), lyophilized, 1,000ng/mL upon reconstitution
Phosphate Buffered Saline Concentrate (PBS)
Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)
Tetramethylbenzidine (TMB) Substrate Solution
96 Well Plate included with product Cat. #’s ending in suffixes -22, -76, -86

Description

Description

Human IgE Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications

Specifications

ELISA
20 x 96 tests
Plasma, serum
Human
Immunoglobulin E
96-well plates
7.8ng/mL
10μL
IgE
Documents

For Research Use Only