Please login to your online account to display your discounted pricing

eBiosciencetrade; Human IgA Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human IgA.

$665.55 - $4,197.75

Specifications

For Use With (Application) ELISA
Sample Size 20μL (1:10,000 prediluted)
Synonym Immunoglobulin A
Target IgA
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IgA

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125317

 
ebioscience
LSBMS2096CE
96 tests Each for $665.55

501125319

 
ebioscience
LSBMS2096TENCE
10 x 96 tests Each for $4,197.75
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 20μL (1:10,000 prediluted)
Synonym Immunoglobulin A
Target IgA
Conjugate Biotin
Sample Type Serum, plasma
Sensitivity 0.03ng/mL
Storage Requirements 2° to 8°C
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.