missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Human IgA ELISA Ready-SET-Go!™ Kit

For quantitative detection of human IgA.

Manufacturer:  Invitrogen™ LS885060077

 View more versions of this product

Catalog No. 501125150



Includes: Capture Antibody Concentrate
Detection Antibody Concentrate
human IgA isotype control (standard), lyophilized, 50ng/mL upon reconstitution
Phosphate Buffered Saline Concentrate (PBS)
Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)
Tetramethylbenzidine (TMB) Substrate Solution
96 Well Plate included with product Cat. #’s ending in suffixes -22, -76, -86

Description

Description

Human IgA Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications

Specifications

ELISA
1.6ng/mL
20μL (prediluted 1:10,000)
IgA
20 x 96 tests
Plasma, serum
Human
Immunoglobulin A
Documents

For Research Use Only