Learn More
Invitrogen™ IFN omega Human ELISA Kit
For the quantitative detection of human IFN-omega.
Supplier: Invitrogen™ BMS233TEN
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human IFN-omega
Biotin-Conjugate anti-human IFN-omega monoclonal antibody
Streptavidin-HRP
Human IFN-omega Standard, 6ng/mL
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Description
The Human Interferon omega (Hu IFNβ) ELISA quantitates Hu IFNβ in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IFNβ. Principle of the method The Human IFNβ solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The protein encoded by this gene is produced by macrophages and has antiviral activity.Specifications
P05000 | |
1.5 pg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
IFNW1 | |
4.9% | |
HRP | |
RUO | |
2°C to 8°C | |
Human | |
3 hr. 10 min. |
4.7-300 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
ELISA | |
3467 | |
6.8% | |
Pre-coated 96 well plate, Adhesive Plate Covers, Sample Diluent, Assay Buffer concentrate, Assay Buffer concentrate, Biotinylated Detection Antibody, Wash Buffer, Lysis Buffer, Lysis Buffer, Chromogen, Stop Solution | |
10 x 96 Tests | |
Plasma, 25 μL; Serum, 25 μL; Supernatant, 25 μL | |
IFNWP2, Interferon alpha-II-1, Interferon omega-1 | |
1 hr. 20 min. |
For Research Use Only