Please login to your online account to display your discounted pricing

eBiosciencetrade; Human IFN-omega Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human IFN-omega.

$665.55 - $4,334.05

Specifications

For Use With (Application) ELISA
Sample Size 25μL
Synonym IFNWP2, Interferon alpha-II-1, Interferon omega-1
Target IFN omega
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IFN-omega
Biotin-Conjugate anti-human IFN-omega monoclonal antibody
Streptavidin-HRP
Human IFN-omega Standard, 6000pg/mL
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125395

 
ebioscience
LSBMS233CE
96 tests Each for $665.55

501125396

 
ebioscience
LSBMS233TENCE
10 x 96 tests Each for $4,334.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 25μL
Synonym IFNWP2, Interferon alpha-II-1, Interferon omega-1
Target IFN omega
Conjugate Biotin
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Sensitivity 1.5pg/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.