Please login to your online account to display your discounted pricing

eBioscience™ Human IFN gamma, Recombinant Protein, Carrier-Free

Human Recombinant Protein

$303.05 - $839.55

Specifications

Name IFN gamma
Recombinant Yes
Conjugate Unlabeled
Purity >98%
Purification Method SDS-PAGE
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

509202

 
ebioscience™
34831982
100μg Each for $303.05

509203

 
ebioscience™
34831985
500μg Each for $839.55
Description & Specifications

Specifications

Name IFN gamma
Recombinant Yes
Conjugate Unlabeled
Purity >98%
Purification Method SDS-PAGE
Source E. coli
For Use With (Application) Cytokine Bioassays
Molecular Weight The methionylated N-terminal protein migrates as a 16kDa polypeptide on SDS-PAGE.
Storage Requirements Store at less than or equal to -70°C.

Human IFN gamma is an ∽20 kDa factor produced by activated T, B and NK cells and is an anti-viral and anti-parasitic cytokine. IFN gamma in synergy with other cytokines, such as TNF alpha, inhibits proliferation of normal and transformed cells. Immunomodulatory effects of IFN gamma are exerted on a wide range of cell types expressing the high affinity receptors for IFN-γ. Glycosylation of IFN gamma does not affect its biological activity.