Human IFN-gamma Quantikine ELISA Kit, R&D Systems
a 4.5 hour solid phase ELISA designed to measure IFN-gamma levels in cell culture supernates, serum, and plasma.
Manufacturer:  R & D Systems RDSDIF50
 View more versions of this product
Catalog No. DIF50
Specifications
Description
Kit, ELISA; R&D Systems; Human IFN-gamma Quantikine ELISA Kit; One 96-well plate; 1 Kit; Sufficient materials to run ELISAs on at least one 96-well plate
It contains E. coli-expressed recombinant human IFN-gamma and antibodies raised against the recombinant factor. Results obtained for naturally occurring human IFN-gamma samples showed linear curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for natural human IFN-gamma
Description & Specifications
Specifications
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
3458
1 Plate
8pg/mL
Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL)
Natural and recombinant human IFN-gamma
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates, Serum, EDTA Plasma
Human
Sufficient materials to run ELISAs on at least one 96-well plate