Please login to your online account to display your discounted pricing

Human IFN-gamma Quantikine ELISA Kit, R&D Systems

$611.21 - $2,922.08

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 3458
Sample Type Cell Culture Supernates, Serum, EDTA Plasma
Sample Volume Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DIF50

 
r & d systems
RDSDIF50
1 Plate Each for $611.21

SIF50

 
r & d systems
RDSSIF50
6 Plates Each for $2,922.08
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 3458
Sample Type Cell Culture Supernates, Serum, EDTA Plasma
Sample Volume Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL)
Species Human
Specificity Natural and recombinant human IFN-gamma

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL)
Sensitivity: 8pg/mL
Specificity: Natural and recombinant human IFN-gamma
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.