missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human IFN-gamma DuoSet, R&D Systems

contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant Human IFN-gamma

Manufacturer:  R & D Systems RDSDY28505

 View more versions of this product

Catalog No. DY28505


 

Specifications

Description

Kit, ELISA; R&D Systems; Human IFN-gamma DuoSet; Five 96-well plates; 1 Kit; Sufficient materials to run ELISAs on at least five 96-well plates

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs
  • Economical alternative to complete kits
Description & Specifications

Specifications

ELISA
3458
5 Plates
Human
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit
1 Kit