Please login to your online account to display your discounted pricing

eBioscience™ Human IFN beta, Recombinant Protein, Carrier-Free

Human Recombinant Protein

$346.55 - $1,260.05

Specifications

Structural Form CHO cells (amino acids Met22-Asn187; Accession # AAC41702.1)
Purity >95%
Purification Method SDS-PAGE and HPLC
Molecular Weight 20kDa
Source CHO Cells
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123521

 
ebioscience™
LS34869982
100μg Each for $1,260.05

501129116

 
ebioscience™
LS34869964
20μg Each for $346.55
Description & Specifications

Specifications

Structural Form CHO cells (amino acids Met22-Asn187; Accession # AAC41702.1)
Purity >95%
Purification Method SDS-PAGE and HPLC
Molecular Weight 20kDa
Source CHO Cells
Endotoxin Concentration Less than 0.1ng/μg cytokine as determined by the LAL assay
Formulation Lyophilized from 10mM sodium citrate, 100mM sodium chloride, and 10mM sucrose
Biological Activity The ED50 of this protein, as measured by stimulation of human TF-1 cell proliferation, is less than or equal to 0.1ng/mL, which corresponds to a specific activity of at least 1 x 107 units/mg.
For Use With (Application) Functional Assays
Conjugate Unlabeled
Regulatory Status RUO
Cross Reactivity Human
Name IFN beta
Recombinant Yes

Human interferon beta is a glycosylated member of the type I family of interferons, which also includes IFN alpha. Human IFN alpha and beta share 50% amino acid sequence homology and bind the IFN a/b receptor (IFNAR), which consists of two subunits IFNAR1 and IFNAR2, with different affinities. Binding of IFN to its receptor activates signal transduction, including the JAK/STAT and p38 MAPK pathways. Many studies have established IFN beta as a mediator of anti-viral, anti-proliferative, and anti-inflammatory functions, as well as an effective treatment for multiple sclerosis.