Please login to your online account to display your discounted pricing

Human IFN-alpha Multi-Subtype ELISA Kit, R&D Systems

$879.18 - $3,405.70

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 3440
Sample Type Cell Culture Supernates
Sample Volume Cell Culture Supernates (100μL)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

411052

 
r & d systems
RDS411052
5 Plates Each for $3,405.70

411051

 
r & d systems
RDS411051
1 Plate Each for $879.18
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 3440
Sample Type Cell Culture Supernates
Sample Volume Cell Culture Supernates (100μL)
Sensitivity 12.5pg/mL
Species Human
Sufficient For Sufficient materials to run ELISAs on at least one 96-well plate

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (100μL)
Sensitivity: 12.5pg/mL
Specificity: Human IFN-α
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.