missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Human IFN alpha ELISA Kits (for Supernatant)

Measure levels of human interferon alpha (IFNa) in human cell and tissue culture media supernatant samples using 96-well plates.

$2,856.50 - $2,856.50

Specifications

Assay Range High Sensitivity: 12.5 to 500pg/mL; Extended Range: 156 to 5000pg/mL
Description Human IFN alpha ELISA Kit (Supernatant)
Format Colorimetric ELISA (TMB substrate) in clear 96-well plate
Kit Contents Antibody Concentrate, 1 vial,Antibody Concentrate, 1 vial
Measurement Absorbance at 450nm
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PI411002 Thermo Scientific™
PI411002
5 plate kit Each for $2,856.50
Specifications

Description

Assay IFN-a (human interferon alpha) levels in human cell and tissue culture media (supernatants) with these Thermo Scientific™ Human IFN alpha ELISA Kits (for Supernatant).

These human IFN alpha ELISA kits use pre-titered, matched pairs of coating and detection antibodies to accurately measure interferon alpha (IFN-α) in samples at 100μL per well. These IFN-α kits are validated for use with human cell and tissue culture media and are optimized for both high-sensitivity and extended assay ranges. Also included are calibrated recombinant protein standard, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Human interferon alpha (human IFN-α, IFN-alpha)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well plate

  • Measurement: Absorbance at 450nm

  • Assay Range: 12.5 to 500pg/mL (high sensitivity); 156 to 5000pg/mL (extended range)

  • Sensitivity: Undetermined

  • Precision: Intra-assay CV ≤ 8%; inter-assay CV ≤ 8%

  • Sample Types: Cell culture supernatant, tissue culture media

  • Total Assay Time: 3.25 hours

  • Sample Size: 100μL

  • Specificity: No significant cross-reaction with human IFN-β, IFN-γ or IFN-ω; mouse or rat IFN-α, IFN-β or IFN-γ; bovine IFN-τ

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, substrates, and stop solution.

Description & Specifications

Specifications

High Sensitivity: 12.5 to 500pg/mL; Extended Range: 156 to 5000pg/mL
Colorimetric ELISA (TMB substrate) in clear 96-well plate
Absorbance at 450nm
100µL
Undetermined
Human interferon alpha (human IFN-a, IFN-alpha)
Human IFN alpha ELISA Kit (Supernatant)
Antibody Concentrate, 1 vial,Antibody Concentrate, 1 vial
Intra-assay CV ≤ 8%; inter-assay CV ≤ 8%
Cell culture supernatant, tissue culture media
No significant cross-reaction with human IFN-ß, IFN-g or IFN-w; mouse or rat IFN-a, IFN-ß or IFN-g; bovine IFN-t
3.25 hr.