missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Human HSP90 alpha Platinum ELISA Kit

For the quantitative detection of human HSP90 alpha.

Manufacturer:  Invitrogen™ LSBMS2090TEN

 View more versions of this product

Catalog No. 501125299



Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human HSP90 alpha
Biotin-Conjugate anti-human HSP90 alpha monoclonal antibody
Streptavidin-HRP
Human HSP90 alpha Standard lyophilized, 20ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. There are 2 major cytosolic HSP90 proteins, HSP90AA1, an inducible form, and HSP90AB1 (MIM 140572), a constitutive form. Other HSP90 proteins are found in endoplasmic reticulum (HSP90B1; MIM 191175) and mitochondria (TRAP1; MIM 606219) (Chen et al., 2005 [PubMed 16269234]).
Specifications

Specifications

P07900
Biotin
3320
Pre-coated 96 well plate, Adhesive Plate Covers, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Controls
10 x 96 tests
0.03 ng/mL
100μL (1:25 prediluted)
Human
1 hr 20 min
Human
Heat Shock Protein 90 alpha
8.6%
0.156-10 ng/mL
ELISA
Colorimetric Microplate Reader
ELISA
RUO
Cell Lysate , Plasma, Serum
Cell Lysate (100μL), Plasma (10μL), Serum (10μL)
2-8°C
4 hr
HSP90AA1
10%
HRP
Documents