Please login to your online account to display your discounted pricing

eBiosciencetrade; Human HSP90 alpha Platinum ELISA Kit

For the quantitative detection of human HSP90 alpha.

$651.05 - $4,189.05

Specifications

For Use With (Application) ELISA
Sample Size 100μL (1:25 prediluted)
Synonym Heat Shock Protein 90 alpha
Target HSP90 alpha
Sample Type Cell lysate, serum and plasma (citrate, heparin)
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human HSP90 alpha
Biotin-Conjugate anti-human HSP90 alpha monoclonal antibody
Streptavidin-HRP
Human HSP90 alpha Standard lyophilized, 20ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125299

 
ebioscience
LSBMS2090TEN
10 x 96 tests Each for $4,189.05

501128684

 
ebioscience
LSBMS2090
96 tests Each for $651.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 100μL (1:25 prediluted)
Synonym Heat Shock Protein 90 alpha
Target HSP90 alpha
Sample Type Cell lysate, serum and plasma (citrate, heparin)
Sensitivity 0.03ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.