Please login to your online account to display your discounted pricing

eBiosciencetrade; Human HSP27 Platinum ELISA Kit

For the quantitative detection of human HSP27.

$651.05 - $4,189.05

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym Heat shock protein 27, heat shock protein beta-1, germ cell-dependent phosphoprotein-1
Target hsp27
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human HSP27
Biotin-Conjugate anti-human HSP27 monoclonal antibody
Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125285

 
ebioscience
LSBMS2086
96 tests Each for $651.05

501125286

 
ebioscience
LSBMS2086FIVE
5 x 96 tests Each for $2,159.05

501125287

 
ebioscience
LSBMS2086TEN
10 x 96 tests Each for $4,189.05

501125288

 
ebioscience
LSBMS2086TWO
2 x 96 tests Each for $941.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym Heat shock protein 27, heat shock protein beta-1, germ cell-dependent phosphoprotein-1
Target hsp27
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Sensitivity 10.9pg/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.