Invitrogen™ eBioscience™ Human HSP27 Platinum ELISA Kit
For the quantitative detection of human HSP27.
Manufacturer:  Invitrogen™ LSBMS2086TWO
 View more versions of this product
Catalog No. 501125288
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human HSP27
Biotin-Conjugate anti-human HSP27 monoclonal antibody
Description
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
ELISA
10.9pg/mL
10μL
hsp27
2 x 96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Human
Heat shock protein 27, heat shock protein beta-1, germ cell-dependent phosphoprotein-1
Documents