missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human HMW Adiponectin/Acrp30 Quantikine ELISA Kit, R&D Systems

$676.55 - $3,247.17

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 9370
Sample Volume Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL)
Sensitivity 0.989ng/mL
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
DHWAD0 R & D Systems
RDSDHWAD0
1 Plate Sufficient materials to run ELISAs on at least one 96-well plate Each for $676.55
SHWAD0 R & D Systems
RDSSHWAD0
6 Plates Sufficient materials to run ELISAs on at least six 96-well plates Each for $3,247.17
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL)
Sensitivity: 0.989ng/mL
Specificity: Natural and recombinant human high molecular weight Adiponectin
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
9370
0.989ng/mL
Solid Phase Sandwich ELISA
Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL)
Human