Please login to your online account to display your discounted pricing

Human HGF Quantikine ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSDHG00

 View more versions of this product

Catalog No. DHG00

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 3082
Product Type Solid Phase Sandwich ELISA
Packaging Quantity 1 Plate
Quantity 1 Kit
Sensitivity 40pg/mL
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Citrate Plasma
Sample Volume Cell Culture Supernates (50μL), Serum (50μL), EDTA Plasma (50μL), Citrate Plasma (50μL)
Species Human
Sufficient For Sufficient materials to run ELISAs on at least one 96-well plate