Please login to your online account to display your discounted pricing

Human HGF Quantikine ELISA Kit, R&D Systems

$663.51 - $3,180.69

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 3082
Sample Volume Cell Culture Supernates (50μL), Serum (50μL), EDTA Plasma (50μL), Citrate Plasma (50μL)
Sensitivity 40pg/mL
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity    

DHG00

 
r & d systems
RDSDHG00
1 Plate Sufficient materials to run ELISAs on at least one 96-well plate Each for $663.51

SHG00

 
r & d systems
RDSSHG00
6 Plates Sufficient materials to run ELISAs on at least six 96-well plates Each for $3,180.69
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 3082
Sample Volume Cell Culture Supernates (50μL), Serum (50μL), EDTA Plasma (50μL), Citrate Plasma (50μL)
Sensitivity 40pg/mL
Species Human

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL), Serum (50μL), EDTA Plasma (50μL), Citrate Plasma (50μL)
Sensitivity: 40pg/mL
Specificity: Natural and recombinant human HGF
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.