missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HEXA Control Fragment Recombinant Protein

Catalog No. RP91303
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP91303 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP91303 Supplier Invitrogen™ Supplier No. RP91303
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-139763 (PA5-139763. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes the alpha subunit of the lysosomal enzyme beta-hexosaminidase that, together with the cofactor GM2 activator protein, catalyzes the degradation of the ganglioside GM2, and other molecules containing terminal N-acetyl hexosamines. Beta-hexosaminidase is composed of two subunits, alpha and beta, which are encoded by separate genes. Both beta-hexosaminidase alpha and beta subunits are members of family 20 of glycosyl hydrolases. Mutations in the alpha or beta subunit genes lead to an accumulation of GM2 ganglioside in neurons and neurodegenerative disorders termed the GM2 gangliosidoses. Alpha subunit gene mutations lead to Tay-Sachs disease (GM2-gangliosidosis type I).

Specifications

Accession Number P06865
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3073
Name Human HEXA Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias beta-hexosaminidase alpha-subunit; beta-hexosaminidase subunit alpha; Beta-hexosaminidase subunit beta; Beta-hexosaminidase subunit beta chain A; Beta-hexosaminidase subunit beta chain B; Beta-N-acetylhexosaminidase subunit alpha; beta-N-acetylhexosaminidase subunit beta; Cervical cancer proto-oncogene 7 protein; EC 3.2.1; EC 3.2.1.52; ENC-1 AS; epididymis luminal protein 248; epididymis secretory protein Li 111; glycosidase; HCC7; HCC-7; HEL-248; HEL-S-111; Hex-1; HEXA; Hexb; hexosaminidase A; hexosaminidase A (alpha polypeptide); hexosaminidase A alpha; hexosaminidase B; hexosaminidase B (beta polypeptide); hexosaminidase subunit A; hexosaminidase subunit alpha; hexosaminidase subunit B; hexosaminidase subunit beta; Hexose aminidase A (alpha polypeptide); MGC99608; N-acetyl-beta-glucosaminidase subunit alpha; N-acetyl-beta-glucosaminidase subunit beta; TSD
Common Name HEXA
Gene Symbol HEXA
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LESFYIQTLLDIVSSYGKGYVVWQEVFDNKVKIQPDTIIQVWREDIPVNYMKELELVTKAGFRALLSAPWYLNRISYGPDWKDFYVVEPLAFE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less