Please login to your online account to display your discounted pricing

eBioscience™ Human Granzyme B Platinum ELISA Kit

For the quantitative detection of human Granzyme B.

$520.84 - $2,932.34

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Granzyme 2, Fragmentin 2, Human lymphocyte protein
Target Granzyme B
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human Granzyme B
Biotin-Conjugate anti-human Granzyme B monoclonal antibody
Streptavidin-HRP
Human Granzyme B Standard lyophilized, 960pg/mL upon reconstitution
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Dilution Buffer Concentrate 5x
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5017910

 
life technologies
LSBMS2027
96 tests Each for $520.84

5017911

 
life technologies
LSBMS2027TEN
10 x 96 tests Each for $2,932.34
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Granzyme 2, Fragmentin 2, Human lymphocyte protein
Target Granzyme B
Conjugate Biotin
Sample Type Cell culture supernatant, serum
Sensitivity 0.2pg/mL
Species Human

  • Percentages of the Granzyme A and B positive CTLs can be determined by flow cytometry and immunocytochemical methods for many disorders
  • Not all granzymes enter the target cell, part of them also 'leak' in to the peripheral blood and other biological fluids
  • Detectable amounts of granzymes have been found to circulate in healthy volunteers
  • These soluble granzymes can be measured by ELISAs