Please login to your online account to display your discounted pricing

eBioscience™ Human GM-CSF, Recombinant Protein, Carrier-Free

Human Recombinant Protein

$1,115.05 - $2,739.05

Specifications

Name GM-CSF
Recombinant Yes
Conjugate Unlabeled
Purity ≥95%
Purification Method SDS-PAGE
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

509215

 
life technologies
LS34833985
500μg Each for $2,739.05

509214

 
life technologies
LS34833982
100μg Each for $1,115.05
Description & Specifications

Specifications

Name GM-CSF
Recombinant Yes
Conjugate Unlabeled
Purity ≥95%
Purification Method SDS-PAGE
Source E. coli
For Use With (Application) Cytokine Bioassays
Molecular Weight The protein is not methionylated at the N-terminal and has a predicted molecular mass of 14,478. The DTT reduced protein migrates as a 14kDa polypeptide on SDS-PAGE. The non-reduced protein migrates as a 13kDa polypeptide.
Storage Requirements Store at less than or equal to -70°C.

Human GM-CSF (Granulocyte/Monocyte-Colony Stimulating Factor) is a differentially glycosylated factor produced mainly by activated T cells and macrophages. Endothelial cells and fibroblasts can also produce GM-CSF after exposure to TNF alpha, IL-1, IL-2 and IFN gamma. GM-CSF is found associated with extracellular matrix and in membrane-bound formats too. GM-CSF stimulates proliferation, activation and differentiation of macrophages and granulocytes and their progenitors.