Invitrogen™ eBioscience™ Human GM-CSF ELISA Ready-SET-Go!™ Kit, 2nd Generation
Specifically engineered for accurate and precise measurement protein levels from samples including serum, plasma, and cell culture supernatants. This second generation kit has increased sensitivity, with a standard curve range from 6-750pg/mL.
Manufacturer:  Invitrogen™ LS88833777
 View more versions of this product
Catalog No. 501125222
Alert:
Contains Sodium azide
Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
10X Coating Buffer . Buffer for plating the Capture Antibody
5X ELISA/ELISPOT Diluent . Buffer for blocking and diluting the Detection Antibody and Enzyme
Enzyme. Pre-titrated Avidin-HRP
Substrate. 1X TMB Solution
Description
Description
Human GM-CSF Ready-SET-Go! (2nd Generation) ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications
Specifications
ELISA
6pg/mL
2° to 8°C except standard which should be stored at ≤−70°C
Granulocyte Macrophage Colony-Stimulating Factor, CSF2
20 x 96 tests
Human
GM-CSF