Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Granulocyte Macrophage Colony Stimulating Factor Recombinant, Thermo Scientific™

Granulocyte Macrophage Colony Stimulating Factor Recombinant protein SGMCSF detects Granulocyte Macrophage Colony Stimulating Factor in Not Applicable samples, and is validated for Control, ELISA and WB

$651.05 - $2,436.00

Products
Catalog Number Mfr. No. Sufficient For Price Quantity    

ENEHGMCSF

 
thermo scientific™
PIEHGMCSF
96 microplate wells Each for $651.05

ENEHGMCSF5

 
thermo scientific™
PIEHGMCSF5
5 x 96 microplate wells Pack of 5 Each for $2,436.00
Description & Specifications

Granulocyte Macrophage Colony Stimulating Factor Recombinant protein SGMCSF detects Granulocyte Macrophage Colony Stimulating Factor in Not Applicable samples, and is validated for Control, ELISA and WB

BRS3 expression has been reported in lung (normal and cancer), nasal mucosa, placenta, and uterus. ESTs have been isolated from normal brain and kidney cancer libraries. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signal
Host Species: E. Coli
Cross Reactivity: Not Applicable
Formulation: 1 Vial purified Protein (1.5 ng/ml) in 4% BSA and no preservative;

PA5-32680 detects BRS3 in Human samples. PA5-32680 has been successfully used in ELISA, Immunohistochemistry and Immunohistochemistry (Paraffin) procedures. PA5-32680 The immunogen is Synthetic 19 amino acid peptide from N-terminal extracellular domain of human BRS3..