Please login to your online account to display your discounted pricing

eBioscience™ Human Glial Cell Derived Neurotrophic Factor (GDNF), Recombinant Protein

Human Recombinant Protein

$201.55 - $419.05

Specifications

Name Glial Cell Derived Neurotrophic Factor (GDNF)
Recombinant Yes
Conjugate Unlabeled
Purity >98%
Purification Method SDS-PAGE
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

5018522

 
life technologies
LS14850662
5μg Each for $201.55

5018523

 
life technologies
LS14850680
25μg Each for $419.05
Description & Specifications

Specifications

Name Glial Cell Derived Neurotrophic Factor (GDNF)
Recombinant Yes
Conjugate Unlabeled
Purity >98%
Purification Method SDS-PAGE
Source E. coli
For Use With (Application) Cytokine Bioassays
Molecular Weight 30kDa (homodimer)
Storage Requirements Store at less than or equal to -70°C.

Glial Cell derived Neurotrophic Factor (GDNF), a member of the TGFß superfamily, is a neurotrophic factor that promotes the survival of various neuronal populations in both the central and peripheral nervous systems during their development. Neuronal subpopulations affected by GDNF include motor neurons, midbrain dopaminergic neurons and Purkinje cells. GDNF is conserved across mammalian species with human and rat orthologs sharing approximately 93% amino acid sequence identity. E. coli-produced recombinant human GDNF is a disulfide-linked, non-glycosylated homodimer composed of two 135 amino acid polypeptides.