missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Gemin 4 (aa 95-185) Control Fragment Recombinant Protein

Catalog No. RP105364
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP105364 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP105364 Supplier Invitrogen™ Supplier No. RP105364
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66789 (PA5-66789. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The product of this gene is part of a large complex localized to the cytoplasm, nucleoli, and to discrete nuclear bodies called Gemini bodies. The complex functions in spliceosomal snRNP assembly in the cytoplasm, and regenerates spliceosomes required for pre-mRNA splicing in the nucleus. The encoded protein directly interacts with a DEAD box protein and several spliceosome core proteins. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.

Specifications

Accession Number P57678
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 50628
Name Human Gemin 4 (aa 95-185) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4932415L08Rik; AU015359; component of gems 4; DKFZp434B131; DKFZp434D174; gem (nuclear organelle) associated protein 4; gem nuclear organelle associated protein 4; gem-associated protein 4; GEMIN4; Gemin-4; HC56; HCAP1; HCC-associated protein 1; HHRF-1; p97; RGD1563715
Common Name Gemin 4
Gene Symbol GEMIN4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DLFFSVGNMIPTINHTILFELLKSLEASGLFIQLLMALPTTICHAELERFLEHVTVDTSAEDVAFFLDVWWEVMKHKGHPQDPLLSQFSAM
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less