Please login to your online account to display your discounted pricing

Human FGF basic Quantikine ELISA Kit, R&D Systems

A 4.5 hour solid phase ELISA designed to measure human FGF basic in cell culture supernates, serum, and plasma. It contains E. coli-expressed recombinant human FGF basic and antibodies raised against the recombinant factor.

$649.50 - $3,113.49

Specifications

Cross Reactivity Cross-species reactivity observed with 1 or more species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 2247
Sample Volume Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL)
Sensitivity 3pg/mL
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity    

DFB50

 
r & d systems
RDSDFB50
1 Plate Sufficient materials to run ELISAs on at least one 96-well plate Each for $649.50

SFB50

 
r & d systems
RDSSFB50
6 Plates Sufficient materials to run ELISAs on at least six 96-well plates Each for $3,113.49
Description & Specifications

Specifications

Cross Reactivity Cross-species reactivity observed with 1 or more species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 2247
Sample Volume Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL)
Sensitivity 3pg/mL
Species Human

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL)
Sensitivity: 3pg/mL
Specificity: Natural and recombinant human FGF basic
Cross Reactivity: Cross-species reactivity observed with 1 or more species tested. <0.5% cross-reactivity observed with available related molecules.