Please login to your online account to display your discounted pricing

eBioscience Human Fc Receptor Binding Inhibitor, Functional Grade Purified

Can be used to inhibit the non-specific Fc-gamma receptor (FcγR)-mediated binding of mouse monoclonal antibodies used for flow cytometric analysis of human tissue.

$143.55 - $462.55

Specifications

Host Species Human
Formulation Aqueous buffer, no sodium azide
Storage Requirements 2° to 8°C
For Use With (Application) Flow Cytometric Analysis
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014778

 
ebioscience
16916171
25 tests Each for $143.55

501129027

 
ebioscience
16916173
100 tests Each for $462.55
Description & Specifications

Specifications

Host Species Human
Formulation Aqueous buffer, no sodium azide
Storage Requirements 2° to 8°C
For Use With (Application) Flow Cytometric Analysis

Use in sterile environment