Please login to your online account to display your discounted pricing

eBiosciencetrade; Human Fc epsilon RI Platinum ELISA Kit

For the detection of human Fc epsilon RI

$651.05 - $4,189.05

Specifications

For Use With (Application) ELISA
Sample Size Serum, plasma (EDTA), plasma (citrate) and plasma (heparin)
Synonym FCER1, FceRI, Fc epsilon RI
Target Fc epsilon Receptor 1 alpha (FceRI)
Sensitivity 0.068ng/mL
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human Fc epsilon RI
Detection antibody (chimeric IgE)
HRP-Conjugate anti-human IgE polyclonal antibody
Human Fc epsilon RI Standard lyophilized, 10ng/mL upon reconstitution
Assay Buffer Concentrate 1x
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films
Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

501125336

 
ebioscience
LSBMS2101
Each for $651.05

501125337

 
ebioscience
LSBMS2101TEN
96-well plates 10 x 96 tests Each for $4,189.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size Serum, plasma (EDTA), plasma (citrate) and plasma (heparin)
Synonym FCER1, FceRI, Fc epsilon RI
Target Fc epsilon Receptor 1 alpha (FceRI)
Sensitivity 0.068ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.