Invitrogen™ eBioscience™ Human Fc epsilon RI Platinum ELISA Kit
For the detection of human Fc epsilon RI
Manufacturer:  Invitrogen™ LSBMS2101
 View more versions of this product
Catalog No. 501125336
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human Fc epsilon RI
Detection antibody (chimeric IgE)
HRP-Conjugate anti-human IgE polyclonal antibody
Human Fc epsilon RI Standard lyophilized, 10ng/mL upon reconstitution
Assay Buffer Concentrate 1x
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films
Description
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
ELISA
0.068ng/mL
Human
FCER1, FceRI, Fc epsilon RI
96 tests
Serum, plasma (EDTA), plasma (citrate) and plasma (heparin)
Fc epsilon Receptor 1 alpha (FceRI)
Documents