missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FAM81B (aa 1-74) Control Fragment Recombinant Protein

Catalog No. RP104090
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104090 Supplier Invitrogen™ Supplier No. RP104090
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (31%), Rat (31%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64476 (PA5-64476. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

FAM81B is a protein coding gene.

Specifications

Accession Number Q96LP2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 153643
Name Human FAM81B (aa 1-74) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias FAM81B; family with sequence similarity 81 member B; family with sequence similarity 81, member B; Gm276; Protein FAM81B; RGD1561830
Common Name FAM81B
Gene Symbol FAM81B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MQLQFLGTLASSEKRKKSQRLFFKNIKSTKNKAGKASIMSSDTNVNKSASPTATAEEQPVEPDGPLPGSDNNQE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less