Please login to your online account to display your discounted pricing

eBioscience Human EPO Platinum ELISA Kit, short incubation, for in vitro diagnostic use

For the quantitative detection of human Erythropoietin. Cell culture supernatant, serum and plasma (EDTA, citrate, heparin), were tested with this assay. Erythropoietin (EPO) is a hormone produced by the kidney that promotes the formation of red blood cells in the bone marrow.

$678.60 - $4,334.05

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Erythropoietin, EPO
Target Erythropoietin (EPO)
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human Erythropoietin
Biotin-Conjugate anti-human Erythropoietin monoclonal antibody
Streptavidin-HRP
Human Erythropoietin Standard lyophilized, 200 mIU/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125265

 
ebioscience
LSBMS2035CE
96 tests Each for $678.60

501125266

 
ebioscience
LSBMS2035TENCE
10 x 96 tests Each for $4,334.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Erythropoietin, EPO
Target Erythropoietin (EPO)
Conjugate Biotin
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Sensitivity 0.17 mIU/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.