Please login to your online account to display your discounted pricing

Human Endostatin Quantikine ELISA Kit, R&D Systems

$676.55 - $3,247.17

Specifications

Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 80781
Sample Volume Cell Culture Supernates (50μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL), Saliva (50μL)
Sensitivity 0.063ng/mL
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity    

DNST0

 
r & d systems
RDSDNST0
1 Plate Sufficient materials to run ELISAs on at least one 96-well plate Each for $676.55

SNST0

 
r & d systems
RDSSNST0
6 Plates Sufficient materials to run ELISAs on at least six 96-well plates Each for $3,247.17
Description & Specifications

Specifications

Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 80781
Sample Volume Cell Culture Supernates (50μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL), Saliva (50μL)
Sensitivity 0.063ng/mL
Species Human

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL), Saliva (50μL)
Sensitivity: 0.063ng/mL
Specificity: Natural and recombinant human Endostatin
Cross Reactivity: Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.