Please login to your online account to display your discounted pricing

eBioscience™ Human CXCL10 (IP-10), Recombinant Protein, Carrier-Free

Human Recombinant Protein

$752.55 - $2,275.05

Specifications

Name CXCL10 (IP-10)
Recombinant Yes
Conjugate Unlabeled
Purity >97%
Purification Method SDS-PAGE
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

50245778

 
ebioscience™
34896782
100μg Each for $752.55

50245779

 
ebioscience™
34896785
500μg Each for $2,275.05
Description & Specifications

Specifications

Name CXCL10 (IP-10)
Recombinant Yes
Conjugate Unlabeled
Purity >97%
Purification Method SDS-PAGE
Source E. coli
For Use With (Application) Cytokine Bioassays
Molecular Weight 8.8kDa
Storage Requirements Store at less than or equal to -70°C.

CXCL10, also known as IP-10 (Interferon Inducible Protein-10), is a member of the CXC family of chemokines. It is secreted by monocytes, epithelial cells, and endothelial cells in response to IFN gamma or other pro-inflammatory cytokines and stimuli. CXCL10 signaling is mediated by the g protein-coupled receptor CXCR3, which is expressed on activated T cells and plays an important role in directing the migration of T cells, especially during Th1 responses. In addition to its role as a chemoattractant, CXCL10 is also a potent inhibitor of angiogenesis and exhibits antitumor activity.