Please login to your online account to display your discounted pricing

Human CXCL1/GRO alpha Quantikine ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSDGR00

 View more versions of this product

Catalog No. DGR00

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 2919
Product Type Solid Phase Sandwich ELISA
Packaging Quantity 1 Plate
Quantity 1 Kit
Sensitivity 10pg/mL
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma, Citrate Plasma
Sample Volume Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Species Human
Sufficient For Sufficient materials to run ELISAs on at least one 96-well plate

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Sensitivity: 10pg/mL
Specificity: Natural and recombinant human GRO alpha
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.