Please login to your online account to display your discounted pricing

Human CXCL1/GRO alpha Quantikine ELISA Kit, R&D Systems

$615.61 - $2,943.11

Specifications

Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 2919
Sample Volume Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Sensitivity 10pg/mL
Species Human
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity    

DGR00

 
r & d systems
RDSDGR00
1 Plate Sufficient materials to run ELISAs on at least one 96-well plate Each for $615.61

SGR00

 
r & d systems
RDSSGR00
6 Plates Sufficient materials to run ELISAs on at least six 96-well plates Each for $2,943.11
Description & Specifications

Specifications

Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 2919
Sample Volume Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Sensitivity 10pg/mL
Species Human

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Sensitivity: 10pg/mL
Specificity: Natural and recombinant human GRO alpha
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.