Please login to your online account to display your discounted pricing

eBioscience Human Cu/ZnSOD Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human Cu/ZnSOD.

$665.55 - $4,334.05

Specifications

For Use With (Application) ELISA
Sample Size 10μL (1:20 prediluted)
Synonym Cu/Zn Superoxide Dismutase
Target Cu/ZnSOD
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human Cu/ZnSOD
HRP-Conjugate anti-human Cu/ZnSOD monoclonal antibody
Human Cu/ZnSOD Standard, 5ng/mL
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Phosphate Buffered Saline Concentrate (PBS) 20x
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125376

 
ebioscience
BMS222TENCE
10 x 96 tests Each for $4,334.05

501125375

 
ebioscience
BMS222CE
96 tests Each for $665.55
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 10μL (1:20 prediluted)
Synonym Cu/Zn Superoxide Dismutase
Target Cu/ZnSOD
Conjugate HRP
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin), amniotic fluid, urine, fetal umbilical vein blood
Sensitivity 0.04ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.