missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CGN (aa 74-163) Control Fragment Recombinant Protein

Catalog No. RP94284
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP94284 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP94284 Supplier Invitrogen™ Supplier No. RP94284
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (72%), Rat (72%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55662 (PA5-55662. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Probably plays a role in the formation and regulation of the tight junction (TJ) paracellular permeability barrier. [UniProt]

Specifications

Accession Number Q9P2M7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 57530
Name Human CGN (aa 74-163) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6330408J11Rik; CGN; Cingulin; KIAA1319
Common Name CGN
Gene Symbol CGN
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GGDSFGVQIKGANDQGASGALSSDLELPENPYSQVKGFPAPSQSSTSDEEPGAYWNGKLLRSHSQASLAGPGPVDPSNRSNSMLELAPKV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less