Please login to your online account to display your discounted pricing

Human CD40 Ligand/TNFSF5 Quantikine ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSSCDL40

 View more versions of this product

Catalog No. SCDL40

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 959
Packaging Quantity 6 Plates
Quantity 1 Pk.
Sensitivity 10.1pg/mL
Sample Type Cell Culture Supernates, Serum, Platelet-poor EDTA Plasma, Platelet-poor Heparin Plasma
Sample Volume Cell Culture Supernates (100μL), Serum (20μL), Platelet-poor EDTA Plasma (100μL), Platelet-poor Heparin Plasma (100μL)
Species Human
Sufficient For Sufficient materials to run ELISAs on at least six 96-well plates