Please login to your online account to display your discounted pricing

eBioscience™ Human CD258 (LIGHT) ELISA Ready-SET-Go!™ Kit

Specifically engineered for accurate and precise measurement of protein levels from samples including serum, plasma, and cell culture supernatants. CD258, or LIGHT, is a 29 kd type II transmembrane protein and a member of the TNF ligand superfamily.

$303.05 - $590.37

Specifications

For Use With (Application) ELISA
Synonym TNFSF14, TR2, HVEML, LTg
Target CD258 (LIGHT)
Sensitivity 75pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
View More Specs
Includes:  Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
10X Coating Buffer . Buffer for plating the Capture Antibody
5X ELISA/ELISPOT Diluent . Buffer for blocking and diluting the Detection Antibody and Enzyme
Enzyme. Pre-titrated Avidin-HRP
Substrate. 1X TMB Solution

 Alert

Contains Sodium azide

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

501125192

 
life technologies
LS88725822
96-well plates 2 x 96 tests Each for $303.05

501125193

 
life technologies
LS88725886
96-well plates 10 x 96 tests Each for $590.37

501125194

 
life technologies
LS88725888
Each for $565.72
Description & Specifications

Specifications

For Use With (Application) ELISA
Synonym TNFSF14, TR2, HVEML, LTg
Target CD258 (LIGHT)
Sensitivity 75pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
Species Human

Human CD258 (LIGHT) Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)