missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human CD25/IL-2 R alpha Quantikine ELISA Kit, R&D Systems

A 3.5 hour solid phase ELISA designed to measure IL-2 sR alpha in cell culture supernates, serum, and plasma. It contains NS0-expressed recombinant human IL-2 sR alpha and antibodies raised against the recombinant factor.

$663.51 - $3,180.69

Specifications

Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 3559
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma
Sample Volume Cell Culture Supernates (10μL), Serum (13μL), EDTA Plasma (13μL), Heparin Plasma (13μL)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
DR2A00 R & D Systems
RDSDR2A00
1 Plate Each for $663.51
SR2A00 R & D Systems
RDSSR2A00
6 Plates Each for $3,180.69
Specifications

Description

The biological effects of interleukin 2 (IL-2) are mediated by its binding to a multi-chain cellular receptor complex. The IL-2 receptor (IL-2 R) complex consists of three glycoprotein chains. The alpha chain (IL-2 R alpha/CD25) and beta chain (IL-2 R beta/CD122) act together with the gamma chain (IL-2 R gamma/CD132) to form a high affinity receptor that transduces the signal upon IL-2 binding.

Alone, CD25/IL-2 R alpha binds IL-2 with low affinity, but is unable to transduce a signal. The alpha/beta combination binds IL-2 with intermediate affinity, but is still unable to transmit a signal inside the cell. The beta/gamma complex has intermediate affinity and is capable of signaling if the concentration of IL-2 is relatively high.

A soluble form of CD25/IL-2 R alpha is present in the serum in parallel with its increased cellular expression. There are reports of soluble IL-2 R beta as well. The biologic function of soluble CD25/IL-2 R alpha is unclear because, due to its low affinity, it is expected to be a poor inhibitor of IL-2. CD25/IL-2 R alpha is a precursor type I membrane protein with a signal peptide, an extracellular region, a transmembrane region, and a cytoplasmic domain. Studies suggest a correlation between the levels of CD25/IL-2 R alpha in serum with the onset of rejection in allograft recipients, with autoimmune disease activation in rheumatoid arthritis and systemic lupus erythematosis patients and with the course of some leukemias and lymphomas.

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (10μL), Serum (13μL), EDTA Plasma (13μL), Heparin Plasma (13μL)
Sensitivity: 10pg/mL
Specificity: Natural and recombinant human IL-2 sR alpha
Cross Reactivity: Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
3559
Cell Culture Supernates (10μL), Serum (13μL), EDTA Plasma (13μL), Heparin Plasma (13μL)
Human
Sufficient materials to run ELISAs on at least six 96-well plates
Solid Phase Sandwich ELISA
Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma
10pg/mL
Natural and recombinant human CD25/IL-2 sR alpha