Please login to your online account to display your discounted pricing

Human CD25/IL-2 R alpha Quantikine ELISA Kit, R&D Systems


A 3.5 hour solid phase ELISA designed to measure IL-2 sR alpha in cell culture supernates, serum, and plasma. It contains NS0-expressed recombinant human IL-2 sR alpha and antibodies raised against the recombinant factor.

Manufacturer: r & d systems  RDSSR2A00

 View more versions of this product

Catalog No. SR2A00

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
For Use With (Application) ELISA
Gene ID (Entrez) 3559
Product Type Solid Phase Sandwich ELISA
Packaging Quantity 6 Plates
Quantity 1 Pk.
Sensitivity 10pg/mL
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma
Sample Volume Cell Culture Supernates (10μL), Serum (13μL), EDTA Plasma (13μL), Heparin Plasma (13μL)
Species Human
Specificity Natural and recombinant human CD25/IL-2 sR alpha
Sufficient For Sufficient materials to run ELISAs on at least six 96-well plates