Please login to your online account to display your discounted pricing

eBioscience™ Human CD154 (CD40 Ligand), sCD154 (sCD40 Ligand), Recombinant Protein, Carrier-Free

Human Recombinant Protein

$172.55 - $1,883.55

Specifications

Name CD154 (CD40 Ligand), sCD154 (sCD40 Ligand)
Recombinant Yes
Conjugate Unlabeled
Purity >98%
Purification Method SDS-PAGE and HPLC
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123530

 
ebioscience™
34890263
10μg Each for $172.55

501123531

 
ebioscience™
34890281
50μg Each for $361.05

501129292

 
ebioscience™
34890285
500μg Each for $1,883.55
Description & Specifications

Specifications

Name CD154 (CD40 Ligand), sCD154 (sCD40 Ligand)
Recombinant Yes
Conjugate Unlabeled
Purity >98%
Purification Method SDS-PAGE and HPLC
Source E. coli
For Use With (Application) Functional Assays
Molecular Weight 16.3kDa
Storage Requirements Store lyophilized protein at less than or equal to -20°C. Reconstituted protein solution can be stored with carrier protein (e.g., 0.1% BSA) at less than or equal to -20°C.

CD40 ligand, (CD40L, also known as CD154, TRAP or gp39) is a membrane glycoprotein expressed on activated CD4+ T-cells, NK cells, mast cells, basophils and eosinophils. The CD40-CD40L interaction stimulates B cell immune response which includes cell surface antigen expression, cell cycle activation, Ig isotype switching, Ig secretion and memory generation. The CD40-CD40L interaction also plays important roles in monocyte and dendritic cell activation, T-cell co-stimulation and cytokine production. It has been reported that the CD40-CD40L interaction is involved in the pathogenesis of amyloid pathology in Alzheimer disease.