Please login to your online account to display your discounted pricing

Human CCL4/MIP-1 beta Quantikine ELISA Kit, R&D Systems

$611.21 - $2,922.08

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 6351
Sample Volume Cell Culture Supernates (10μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Sensitivity 4pg/mL (C), 11pg/mL (S, P)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity    

DMB00

 
r & d systems
RDSDMB00
1 Plate Sufficient materials to run ELISAs on at least one 96-well plate Each for $611.21

SMB00

 
r & d systems
RDSSMB00
6 Plates Sufficient materials to run ELISAs on at least six 96-well plates Each for $2,922.08
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 6351
Sample Volume Cell Culture Supernates (10μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Sensitivity 4pg/mL (C), 11pg/mL (S, P)
Species Human

Human MIP-1 beta (macrophage inflammatory protein 1 beta) is the product of several independently cloned cDNAs including Act 2, PAT 744, hH400, G26, HIMAP, HC21, and MAD-5a. MIP-1 beta is a member of the CC or beta chemokine subfamily that was originally purified from the conditioned media of an LPS-stimulated murine macrophage cell line. MIP-1 beta is expressed primarily by T cells, B cells, and monocytes after antigen or mitogen stimulation. The functional receptor for MIP-1 beta has been identified as CCR5.

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (10μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Sensitivity: 4pg/mL (C), 11pg/mL (S, P)
Specificity: Natural and recombinant human MIP-1 beta
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.