Please login to your online account to display your discounted pricing

eBioscience Human CCL4 (MIP-1 beta) ELISA Ready-SET-Go!™ Kit

Specifically engineered for accurate and precise measurement of CCL4 protein levels from samples including serum, plasma, and cell culture supernatants.

$332.05 - $999.05

Specifications

For Use With (Application) ELISA
Synonym C-C motif Chemokine 4, Macrophage Inflammatory Protein 1 beta
Target CCL4 (MIP-1 beta)
Sensitivity 4pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
View More Specs
Includes:  Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
10X Coating Buffer . Buffer for plating the Capture Antibody
5X ELISA/ELISPOT Diluent . Buffer for blocking and diluting the Detection Antibody and Enzyme
Enzyme. Pre-titrated Avidin-HRP
Substrate. 1X TMB Solution

 Alert

Contains Sodium azide

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

501125175

 
ebioscience
LS88703422
96-well plates 2 x 96 tests Each for $332.05

501125176

 
ebioscience
LS88703476
96-well plates 20 x 96 tests Each for $999.05

501125177

 
ebioscience
LS88703477
Each for $941.05

501125178

 
ebioscience
LS88703488
Each for $665.55
Description & Specifications

Specifications

For Use With (Application) ELISA
Synonym C-C motif Chemokine 4, Macrophage Inflammatory Protein 1 beta
Target CCL4 (MIP-1 beta)
Sensitivity 4pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
Species Human

Human CCL4 (MIP-1 beta) Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)