Invitrogen™ eBioscience™ Human CCL3 (MIP-1 alpha) ELISA Ready-SET-Go!™ Kit
Specifically engineered for accurate and precise measurement of CCL3 protein levels from samples including serum, plasma, and cell culture supernatants.
Manufacturer:  Invitrogen™ LS88703577
 View more versions of this product
Catalog No. 501125181
Alert:
Contains Sodium azide
Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
10X Coating Buffer . Buffer for plating the Capture Antibody
5X ELISA/ELISPOT Diluent . Buffer for blocking and diluting the Detection Antibody and Enzyme
Enzyme. Pre-titrated Avidin-HRP
Substrate. 1X TMB Solution
Description
Description
Human CCL3 (MIP-1 alpha) Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers, and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications
Specifications
ELISA
16pg/mL
2° to 8°C except standard which should be stored at ≤−70°C
C-C motif chemokine 3, MIP1A, Macrophage 1nflammatory Protein-1 alpha
20 x 96 tests
Human
CCL3 (MIP-1 alpha)