missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human CCL11/Eotaxin Quantikine ELISA Kit, R&D Systems

$663.51 - $3,180.69

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 6356
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Citrate Plasma
Sample Volume Cell Culture Supernates (50μL), Serum (50μL), EDTA Plasma (50μL), Citrate Plasma (50μL)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
DTX00 R & D Systems
RDSDTX00
1 Plate Sufficient materials to run ELISAs on at least one 96-well plate Each for $663.51
STX00 R & D Systems
RDSSTX00
6 Plates Sufficient materials to run ELISAs on at least six 96-well plates Each for $3,180.69
Specifications

Description

Eotaxin is a potent eosinophil chemoattractant that is a member of the CC chemokine subfamily of inflammatory and immunoregulatory cytokines. At the protein sequence level, mature mouse Eotaxin is approximately 60% identical to mature human and guinea pig Eotaxin. Eotaxin is chemotactic for eosinophils, but not mononuclear cells or neutrophils.

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL), Serum (50μL), EDTA Plasma (50μL), Citrate Plasma (50μL)
Sensitivity: 5pg/mL
Specificity: Natural and recombinant human Eotaxin
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
6356
Cell Culture Supernates (50μL), Serum (50μL), EDTA Plasma (50μL), Citrate Plasma (50μL)
Human
Solid Phase Sandwich ELISA
Cell Culture Supernates, Serum, EDTA Plasma, Citrate Plasma
5pg/mL