Invitrogen™ eBioscience™ Human C5a Platinum ELISA Kit
For the detection of human C5a in serum, plasma (EDTA, citrate, heparin) and cell culture supernatant
Manufacturer:  Invitrogen™ LSBMS2088TEN
 View more versions of this product
Catalog No. 501125292
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human C5a
Description
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
ELISA
0.005ng/mL
Sample volume 10μL for plasma and 5μL for serum (prediluted 1:100 for serum, 1:25 for plasma), 50μL for CCS
C5a
10 x 96 tests
Sample Types serum, plasma (EDTA, citrate, heparin) and cell culture supernatant
Human
complement component 5, C5a, C5D, C5b, ECLZB, CPAMD4
Documents